Diacerein
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
J63775.03
J63775.06
J63775.03EA
162
EUR
J63775.03
J63775.06
Diacerein
Diacerein
CAS No.: 13739-02-1
Molecular Formula: C19H12O8
Formula Weight: 368.29
Physical Form: Powder
Appearance: Pale yellow
Melting Point: 217-218°
MDL No.: MFCD00468030
Applications: Interleukin-1 inhibitor
Formule:
C₁₉H₁₂O₈ Smeltpunt: 217…218 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00468030 CAS nummer: 13739-02-1 EINECS: 237-310-2 Merck Index: 14,2960 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...