Indapamide
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
CAS No.: 26807-65-8
Molecular Formula: C16H16ClN3O3S
Formula Weight: 365.83
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Appearance: White
Melting Point: 160-162°
Solubility: Soluble in ethanol. Insoluble in water
MDL No.: MFCD00079375
Applications: Used as an antihypertensive. Diuretic
Formule:
C₁₆H₁₆ClN₃O₃S Smeltpunt: 160…162 °C |
MDL Nummer:
MFCD00079375 CAS nummer: 26807-65-8 EINECS: 248-012-7 Merck Index: 14,4935 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...