Agarose gel loading dye 6X for electrophoresis, glycerol based
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J63869.ACEA 63.1 EUR
J63869.AC J63869.AD
Agarose gel loading dye 6X for electrophoresis, glycerol based
Kleurstoffen Kleuringen voor elektroforese
Agarose gel loading dye (6X) is used in DNA gel analysis in alkaline conditions.
Used in agarose gel electrophoresis.
Bestelinformatie: Contains 0,25% bromophenol blue, 0,25% xylene cyanol FF and 40% glycerol.