oxolinic acid 98%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Purity: 98%
CAS No.: 14698-29-4
Molecular Formula: C13H11NO5
Formula Weight: 261.23
Storage Temperature: 2°C to 8°C
Flash Point:
Applications: Targets gram negative bacteria
Formule:
C₁₃H₁₁NO₅ Kookpunt: 473 °C Vlampunt: >110 °C (230 °F) |
MDL Nummer:
MFCD00056775 CAS nummer: 14698-29-4 EINECS: 238-750-8 Merck Index: 14,6965 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...