Pyrazinamide 98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synoniemen: Pyrazinecarboxamide, Pyrazine-2-carboxamide, 2-Pyrazinecarboxamide

J66822.09 J66822.14 J66822.22
J66822.09EA 44.8 EUR
J66822.09 J66822.14 J66822.22
Pyrazinamide 98%
Pyrazinamide

Purity: 98% CAS No.: 98-96-4 Molecular Formula: C5H5N3O Formula Weight: 123.11 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Physical Form: Powder Appearance: White Odour: Odourless Melting Point: 189-191° Solubility: Soluble in water Beilstein Reference: 112306 MDL No.: MFCD00006132

Formule: C₅H₅N₃O
Smeltpunt: 189…191 °C
Vlampunt: >110 °C (230 °F)
MDL Nummer: MFCD00006132
CAS nummer: 98-96-4
EINECS: 202-717-6
Merck Index: 14,7956

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR