Silver 0,02 mg/ml supplied in 2 mM sodium citrate, nanoparticles 20 nm (Abs. 405 nm)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J67067.AC J67067.AE
J67067.ACEA 104 EUR
J67067.AC J67067.AE
Silver 0,02 mg/ml supplied in 2 mM sodium citrate, nanoparticles 20 nm (Abs. 405 nm)
Silver
CAS No.: 7440-22-4 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Yellow Odour: Odourless MDL No.: MFCD00003397
Formule: Ag
MW: 107.87 g/mol
Kookpunt: 2212
Smeltpunt: 962
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00003397
CAS nummer: 7440-22-4
Merck Index: 12,08647