Gold 95%, nanoparticles 5 nm OD1 (Abs. 515-520 nm)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J67089.AC J67089.AE
J67089.ACEA 94.1 EUR
J67089.AC J67089.AE
Gold 95%, nanoparticles 5 nm OD1 (Abs. 515-520 nm)
Gold
Purity: 95% CAS No.: 7440-57-5 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Red Odour: Odourless MDL No.: MFCD00003436
Applications: For development of peptide and protein gold conjugates for use in applications such as blotting, microscopy and TEM
Formule: Au
MW: 196.97 g/mol
Kookpunt: 2800
Smeltpunt: 1063
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00003436
CAS nummer: 7440-57-5
Merck Index: 12,04526