Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 100 nm OD1 (Abs. 572 nm)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J67095.AC J67095.AE
J67095.ACEA 98.8 EUR
J67095.AC J67095.AE
Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 100 nm OD1 (Abs. 572 nm)
Gold
CAS No.: 7440-57-5 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Red to pink MDL No.: MFCD00003436
Formule: Au
MW: 196.97 g/mol
Kookpunt: 2800
Smeltpunt: 1063
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00003436
CAS nummer: 7440-57-5
Merck Index: 12,04526