Silver 0,02 mg/ml supplied in 2 mM sodium citrate, nanoparticles 100 nm (Abs. 490 nm)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J67099.AC J67099.AE
J67099.ACEA 102 EUR
J67099.AC J67099.AE
Silver 0,02 mg/ml supplied in 2 mM sodium citrate, nanoparticles 100 nm (Abs. 490 nm)
Silver
CAS No.: 7440-22-4 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Yellow Odour: Odourless MDL No.: MFCD00003397
Applications: An alternative to gold nanoparticles due to lower absorption maximum in the 400-500nm range. Readily binds proteins and other ligands for conjugate and sensor development.
Formule: Ag
MW: 107.87 g/mol
Kookpunt: 2212
Smeltpunt: 962
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00003397
CAS nummer: 7440-22-4
Merck Index: 12,08647