Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 80 nm OD1 (Abs. 553 nm)
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J67109.AC
J67109.AE
J67109.ACEA
103
EUR
J67109.AC
J67109.AE
Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 80 nm OD1 (Abs. 553 nm)
Gold
CAS No.: 7440-57-5
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Sensitivity: Light Sensitive
Appearance: Red to pink
MDL No.: MFCD00003436
Applications: Suitable for adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.
Formule:
Au Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00003436 CAS nummer: 7440-57-5 |