Gold 99%, nanoparticles 10 nm reactant-free OD1 (Abs. 520 nm)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J67238.AC J67238.AE
J67238.ACEA 118 EUR
J67238.AC J67238.AE
Gold 99%, nanoparticles 10 nm reactant-free OD1 (Abs. 520 nm)
Gold
Purity: 99% CAS No.: 7440-57-5 Storage Temperature: 2°C to 8°C Physical Form: Liquid Appearance: Red MDL No.: MFCD00003436
Formule: Au
MW: 196.97 g/mol
Kookpunt: 2800
Smeltpunt: 1063
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00003436
CAS nummer: 7440-57-5
Merck Index: 12,04526