Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 20 nm OD1 (Abs. 524 nm)
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
J67269.ACEA 103 EUR
J67269.AC J67269.AE
Gold supplied in 0.1 mg/ml sodium citrate with stabilizer stabilised, nanoparticles 20 nm OD1 (Abs. 524 nm)
Gold
CAS No.: 7440-57-5 Storage Temperature: 2°C to 8°C Physical Form: Liquid Sensitivity: Light Sensitive Appearance: Red to pink MDL No.: MFCD00003436
Applications: For use in adsorption of thiolated ligands such as oligonucleotides and polyethylene glycols. Also, due to the increased stability, these gold nanoparticles are ideal for use in physical and optical applications.
Formule: Au
MW: 196.97 g/mol
Kookpunt: 2800
Smeltpunt: 1063
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00003436
CAS nummer: 7440-57-5
Merck Index: 12,04526