N-Octyl-β-D-glucopyranoside 98+%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen:
Octylglucopyranoside, OG, (2R,3S,4S,5R,6R)-2-(Hydroxymethyl)-6-(octyloxy)tetrahydro-2H-pyran-3,4,5-triol, n-Octyl β-D-glucopyranoside, 1-O-n-Octyl-β-D-glucopyranoside, Octyl-β-D-glucopyranoside
Purity: 98+%
CAS No.: 29836-26-8
Molecular Formula: C14H28O6
Formula Weight: 292.37
Storage Temperature: -30°C to -10°C
Physical Form: Powder
Sensitivity: Hygroscopic
Appearance: White to off-white
Melting Point: 103-104°
Beilstein Reference: 84118
Optical Rotation: -31° (c=2 in methanol)
MDL No.: MFCD00063288
Formule:
C₁₄H₂₈O₆ MW: 292.37 g/mol Smeltpunt: 103…104 °C |
MDL Nummer:
MFCD00063288 CAS nummer: 29836-26-8 EINECS: 249-887-8 |