Boorzuur ≥99.5% for molecular biology
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Synoniemen:
Boron trihydroxide, Boron hydroxide, Boric acid, Boracic acid
J67419.A1
J67419.A7
J67419.36
J67419.A1EA
102
EUR
J67419.A1
J67419.A7
J67419.36
Boorzuur ≥99.5% for molecular biology
Boorzuur
Purity: 99.5% min
CAS No.: 10043-35-3
Molecular Formula: BH3O3
Formula Weight: 61.83
Physical Form: Powder
Density: 1.435
Appearance: White
Odour: Odourless
pH: 3.8-4.8
Melting Point: ca 185° dec
Beilstein Reference: 1697939
MDL No.: MFCD00011337
Formule:
B(OH)₃ Dichtheid: 1,435 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00011337 CAS nummer: 10043-35-3 EINECS: 233-139-2 Merck Index: 14,1336 |