N-Cyclohexylhydroxylamine hydrochloride 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
L03569.03
L03569.06
L03569.03EA
29.1
EUR
L03569.03
L03569.06
N-Cyclohexylhydroxylamine hydrochloride 98%
N-Cyclohexylhydroxylamine hydrochloride
Purity: 98%
CAS No.: 25100-12-3
Molecular Formula: C6H13NO.HCl
Formula Weight: 151.64
Melting Point: ca 166° dec.
Beilstein Reference: 3672798
MDL No.: MFCD00012565
Formule:
C₆H₁₃NO·HCl Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00012565 CAS nummer: 25100-12-3 EINECS: 246-612-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...