6-Azathymine 98%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L06762.06
L06762.14
L06762.06EA
61.2
EUR
L06762.06
L06762.14
6-Azathymine 98%
6-Azathymine
Purity: 98%
CAS No.: 932-53-6
Molecular Formula: C4H5N3O2
Formula Weight: 127.10
Melting Point: 211-215°
Beilstein Reference: 126863
MDL No.: MFCD00006457
Formule:
C₄H₅N₃O₂ Smeltpunt: 211…215 °C Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00006457 CAS nummer: 932-53-6 EINECS: 213-253-9 Merck Index: 14,903 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...