2-(2-Ethoxyethoxy)ethyl bromide ≥98% stabilised
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Synoniemen:
1-Bromo-2-(2-ethoxyethoxy)ethane
Stabilisatie:
Stabilised with sodium carbonate (Na₂CO₃)
L08522.09
L08522.18
L08522.09EA
65.6
EUR
L08522.09
L08522.18
2-(2-Ethoxyethoxy)ethyl bromide ≥98% stabilised
2-(2-Ethoxyethoxy)ethyl bromide
Stab. with potassium carbonate
Purity: 98%
CAS No.: 54550-36-6
Molecular Formula: CH3CH2OCH2CH2OCH2CH2Br
Formula Weight: 197.08
Refractive Index: 1.4530
Boiling Point: 86-88°/12mm
Beilstein Reference: 1736603
MDL No.: MFCD00051920
Formule:
CH₃CH₂OCH₂CH₂OCH₂CH₂Br Kookpunt: 86…88 °C (12 mmHg) Dichtheid: 1,299 Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00051920 CAS nummer: 54550-36-6 |