4-Hydroxybutyl acrylate 95% stabilised
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Stabilisatie:
Stabilised with 4-methoxyphenol (MEHQ) 500 ppm
Stab. with ca 500ppm 4-methoxyphenol
Purity: 95%
CAS No.: 2478-10-6
Molecular Formula: H2C=CHCO2(CH2)4OH
Formula Weight: 144.17
Flash Point:
Formule:
C₇H₁₂O₃ Kookpunt: ∼230 °C Smeltpunt: –112 °C Dichtheid: 1,041 Vlampunt: >110 °C (230 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00010261 CAS nummer: 2478-10-6 EINECS: 219-606-3 UN: 3082 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...