6-Chlorohexanoyl chloride 95%
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Purity: 95%
CAS No.: 19347-73-0
Molecular Formula: Cl(CH2)5COCl
Formula Weight: 169.05
Flash Point:
Formule:
Cl(CH₂)₅COCl Kookpunt: 90 °C (7 mmHg) Dichtheid: 1,400 Vlampunt: >110 °C (230 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00041436 CAS nummer: 19347-73-0 EINECS: 242-979-9 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...