N-tert-Butyl-2-benzothiazolesulphenamide 97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
L13356.18
L13356.30
L13356.18EA
37.4
EUR
L13356.18
L13356.30
N-tert-Butyl-2-benzothiazolesulphenamide 97%
N-tert-Butyl-2-benzothiazolesulphenamide
Purity: 97%
CAS No.: 95-31-8
Molecular Formula: C11H14N2S2
Formula Weight: 238.38
Flash Point: 165°(329°F)
Melting Point: 105-110°
Beilstein Reference: 158370
MDL No.: MFCD00022873
Formule:
C₁₁H₁₄N₂S₂ MW: 238.38 g/mol Smeltpunt: 105…110 °C Dichtheid: 1.290 Vlampunt: 165 °C (329 °F) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00022873 CAS nummer: 95-31-8 EINECS: 202-409-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...