2-(Dimethylamino)ethyl acrylate 98% stabilised
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
Stabilisatie:
Stabilised with 4-methoxyphenol (MEHQ) 0,1%
L13506.AE
L13506.AP
L13506.AC
L13506.AEEA
79.4
EUR
L13506.AE
L13506.AP
L13506.AC
2-(Dimethylamino)ethyl acrylate 98% stabilised
2-(Dimethylamino)ethyl acrylate
Stab. with ca 0.1% 4-methoxyphenol
Purity: 98%
CAS No.: 2439-35-2
Molecular Formula: C7H13NO2
Formula Weight: 143.19
Storage Temperature: 2°C to 8°C
Flash Point: 62°(144°F)
Refractive Index: 1.4380
Melting Point: -60°
Boiling Point: 64°/12mm
Beilstein Reference: 1099119
UN No.: UN3302
MDL No.: MFCD00038233
Formule:
C₇H₁₃NO₂ Kookpunt: 64 °C (12 mmHg) Smeltpunt: –60 °C Dichtheid: 0,943 Vlampunt: 62 °C (144 °F) Bewaar temperatuur: Koelkast |
MDL Nummer:
MFCD00038233 CAS nummer: 2439-35-2 EINECS: 219-460-0 UN: 3302 ADR: 6.1,II |