N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium tetrafluoroborate 98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: O-(N-Succinimidyl)-N,N,N',N'-tetramethyluronium Tetrafluoroborate, TSTU, 2-(2,5-Dioxopyrrolidin-1-yl)-1,1,3,3-tetramethylisouronium tetrafluoroborate
L13538.06 L13538.14
L13538.06EA 89.7 EUR
L13538.06 L13538.14
N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium tetrafluoroborate 98%
N,N,N',N'-Tetramethyl-O-(N-succinimidyl)uronium tetrafluoroborate
Purity: 98% CAS No.: 105832-38-0 Molecular Formula: C9H16BF4N3O3 Formula Weight: 301.05 Melting Point: 195-200° MDL No.: MFCD00077875
Formule: C₉H₁₆BF₄N₃O₃
MW: 301.05 g/mol
Smeltpunt: 195…200 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00077875
CAS nummer: 105832-38-0