Diisopropyl bromomethylphosphonate 97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
L13763.06
L13763.14
L13763.06EA
74.8
EUR
L13763.06
L13763.14
Diisopropyl bromomethylphosphonate 97%
Diisopropyl bromomethylphosphonate
Purity: 97%
CAS No.: 98432-80-5
Molecular Formula: C7H16BrO3P
Formula Weight: 259.09
Refractive Index: 1.4495
Boiling Point: 80°/1mm
Beilstein Reference: 4740388
UN No.: UN3265
MDL No.: MFCD00236036
Formule:
C₇H₁₆BrO₃P Kookpunt: 80 °C (1 mmHg) Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD00236036 CAS nummer: 98432-80-5 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...