Methacrylic anhydride 94% stabilised
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Methacrylic Anhydride [stabilized with 1,1,3-Tris(3-tert-butyl-4-hydroxy-6-methylphenyl)butane]
Stabilisatie: ∼ 0.2% Stabilised with 2,4-dimethyl-6-tert-butylphenol
L14357.18 L14357.30
L14357.18EA 36.7 EUR
L14357.18 L14357.30
Methacrylic anhydride 94% stabilised
Methacrylic anhydride
Stab. with ca 0.2% 2,4-dimethyl-6-tert-butylphenol
Purity: 94% CAS No.: 760-93-0 Molecular Formula: C8H10O3 Formula Weight: 154.17 Flash Point: 84°(183°F) Refractive Index: 1.4540 Melting Point: -109° Boiling Point: 197° Beilstein Reference: 1761982 UN No.: UN3265 MDL No.: MFCD00008586
Formule: C₈H₁₀O₃
MW: 154.165 g/mol
Kookpunt: 197 °C
Smeltpunt: –109 °C
Dichtheid: 1.033
Vlampunt: 84 °C (183 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00008586
CAS nummer: 760-93-0
EINECS: 212-084-8
UN: 3265
ADR: 8 II