4-Methoxy-2-methylphenylhydrazine hydrochloride 96%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
L14810.03
L14810.06
L14810.03EA
33.7
EUR
L14810.03
L14810.06
4-Methoxy-2-methylphenylhydrazine hydrochloride 96%
4-Methoxy-2-methylphenylhydrazine hydrochloride
Purity: 96%
CAS No.: 93048-16-9
Molecular Formula: C8H12N2O.HCl
Formula Weight: 188.66
Melting Point: ca 110° dec.
Beilstein Reference: 7227704
UN No.: UN2811
MDL No.: MFCD03426021
Formule:
C₈H₁₂N₂O·HCl Bewaar temperatuur: Kamertemperatuur |
MDL Nummer:
MFCD03426021 CAS nummer: 93048-16-9 ADR: 6.1,III |