2,3-Dibromofuran 97% stabilised
Leverancier: Thermo Fisher ScientificTotal Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Waarschuwing
|
Stabilisatie:
Stabilised with calcium carbonate 0,5%
Stab. with 0.5% calcium carbonate
Purity: 97%
CAS No.: 30544-34-4
Molecular Formula: C4H2Br2O
Formula Weight: 225.87
Storage Temperature: -30°C to -10°C
Refractive Index: 1.5470
Boiling Point: 75-78°/75mm
Beilstein Reference: 107511
MDL No.: MFCD01074839
Formule:
C₄H₂Br₂O Kookpunt: 75…78 °C (75 mmHg) Dichtheid: 2,000 Bewaar temperatuur: Vriezer |
MDL Nummer:
MFCD01074839 CAS nummer: 30544-34-4 |