4,4,5,5-Tetramethyl-1,3,2-dioxaborolane 97%
Leverancier: Thermo Fisher Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Gevaar
|
L17558.06
L17558.14
L17558.06EA
54.8
EUR
L17558.06
L17558.14
4,4,5,5-Tetramethyl-1,3,2-dioxaborolane 97%
4,4,5,5-Tetramethyl-1,3,2-dioxaborolane
Purity: 97%
CAS No.: 25015-63-8
Molecular Formula: C6H13BO2
Formula Weight: 127.98
Storage Temperature: 2°C to 8°C
Flash Point: 5°(41°F)
Refractive Index: 1.3970
Boiling Point: 42-43°/50mm
Beilstein Reference: 7802871
UN No.: UN3399
MDL No.: MFCD00674030
Formule:
C₆H₁₃BO₂ Kookpunt: 42…43 °C (50 mmHg) Dichtheid: 0,882 Vlampunt: 5 °C (41 °F) |
MDL Nummer:
MFCD00674030 CAS nummer: 25015-63-8 ADR: 4.3,II |