N-Methyl-N-octylamine ≥98%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
L18639.14 L18639.06
L18639.14EA 179 EUR
L18639.14 L18639.06
N-Methyl-N-octylamine ≥98%
N-Methyl-N-octylamine
Purity: 98% CAS No.: 2439-54-5 Molecular Formula: C9H21N Formula Weight: 143.27 Flash Point: 63°(145°F) Refractive Index: 1.4270 Boiling Point: 68°/8mm UN No.: UN2735 MDL No.: MFCD00048927
Formule: C₉H₂₁N
Kookpunt: 68 °C (8 mmHg)
Dichtheid: 0,790
Vlampunt: 63 °C (145 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00048927
CAS nummer: 2439-54-5
EINECS: 219-462-1
UN: 2735
ADR: 8,II