1-Phenyl-2-pyrrolidone 99%
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
L14660.22 L14660.06 L14660.14
L14660.22EA 247 EUR
L14660.22 L14660.06 L14660.14
1-Phenyl-2-pyrrolidone 99%
1-Phenyl-2-pyrrolidone
Odour: Odourless Melting Point: 63-68° Boiling Point: 123°/0.2mm Beilstein Reference: 130685
Formule: C₁₀H₁₁NO
Kookpunt: 123 °C (0,2 mmHg)
Smeltpunt: 63…68 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003192
CAS nummer: 4641-57-0
EINECS: 225-069-6
ADR: 8,I
Merck Index: 13,01337
Order Now
Specificaties
Environmentally Preferable