Polyethylene glycol 8,000 NF
Leverancier: Spectrum Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: PEG 8,000, Ethylene glycol polymer 8000, PEG 8000
PO131-12KG PO131-500GM
SPCMPO131-12KGEA 1040 EUR
SPCMPO131-12KG SPCMPO131-500GM
Polyethylene glycol 8,000 NF
Polyethylene glycol 8,000
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Formule: H(OCH₂CH₂)nOH
MW: 8000 g/mol
Kookpunt: 250
Dichtheid: 1.125 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD01779615
CAS nummer: 25322-68-3
EINECS: 500-038-2
Merck Index: 12,07729
Order Now
Specificaties
Environmentally Preferable