Poloxamer 407 (Pluronic® F-127) NF
Leverancier: Spectrum Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
P1166-2.5KG P1166-45KGBL P1166-500GM P1166-12KG P1166-25KGBL
SPCMP1166-2.5KGEA 648 EUR
SPCMP1166-2.5KG SPCMP1166-45KGBL SPCMP1166-500GM SPCMP1166-12KG SPCMP1166-25KGBL
Poloxamer 407 (Pluronic® F-127) NF
Poloxamer 407 (Pluronic® F-127)
Poloxamer 407, NF is a national formulary approved surfactant that finds utility because of its amphiphilic structure.
  • Amphphilic structure
  • Increases miscibility
  • Controls thickness of personal care products
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities
Formule: HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)aH
MDL Nummer: MFCD00082049
CAS nummer: 9003-11-6