Dextran, M.W. ~ 500,000, powder
Leverancier: Spectrum Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
SPCMD1004-50KGBLEA 0 EUR
SPCMD1004-50KGBL SPCMD1004-100GM SPCMD1004-25GM SPCMD1004-10KGBL SPCMD1004-1KG
Dextran, M.W. ~ 500,000, powder
Dextran
Form: Powder
Formule: (C₆H₁₀O₅)n
MDL Nummer: MFCD00130935
CAS nummer: 9004-54-0
EINECS: 232-677-5