Sodium dodecylbenzenesulphonate, powder
Leverancier: Spectrum Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: Laurylbenzenesulphonic acid Sodium Salt, Alkylbenzenesulphonic acid Sodium Salt, Sodium Alkylbenzenesulphonate, Dodecylbenzenesulphonic acid sodium salt, Sodium Laurylbenzenesulphonate
D1064-12KG D1064-25KGBL D1064-90KGBL
SPCMD1064-12KGEA 951 EUR
SPCMD1064-12KG SPCMD1064-25KGBL SPCMD1064-90KGBL
Sodium dodecylbenzenesulphonate, powder
Sodium dodecylbenzenesulphonate
Form: Powder
Formule: C₁₈H₂₉NaO₃S
Smeltpunt: > 300 °C
Dichtheid: 1 g/cm³ (20 °C)
MDL Nummer: MFCD00011508
CAS nummer: 25155-30-0
EINECS: 246-680-4
Merck Index: 13,08686
Order Now
Specification Test Results
Assay 90% Min.