Diethylene glycol monoethyl ether
Leverancier: Spectrum Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: 2-(2-Ethoxyethoxy)ethanol, Di(ethylene glycol) ethyl ether, Diethylene Glycol Monoethyl Ether
E1022-200LTBL E1022-20LTMT
SPCME1022-200LTBLEA 6030 EUR
SPCME1022-200LTBL SPCME1022-20LTMT
Diethylene glycol monoethyl ether
Diethylene glycol monoethyl ether
BSE/TSE statement (available upon request) Allergen statement (available upon request)
Formule: C₆H₁₄O₃
MW: 134.18 g/mol
Kookpunt: 197
Smeltpunt: –80 °C
Dichtheid: 0.99 g/cm³ (20 °C)
Vlampunt: 93
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002872
CAS nummer: 111-90-0
EINECS: 203-919-7
Bereik: 01-2119475105-42
Merck Index: 13,01809
Order Now
Specification Test Results
Assay 99% Min.