L-Arginine hydrochloride USP
Leverancier: Spectrum Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: H-L-Arg-OH.HCl, (S)-2-Amino-5-guanidinopentanoic acid hydrochloride, (S)-(+)-Arginine HCl, H-Arg-OH·HCl, L-Arginine, hydrochloride, (S)-(+)-Arginine hydrochloride, H-Arg-OH.HCl, L-Arg, L(+)-Arginine monohydrochloride, L(+)-Arginine hydrochloride
A1337-12KGBL A1337-1KG A1337-50KGBL
SPCMA1337-12KGBLEA 3690 EUR
SPCMA1337-12KGBL SPCMA1337-1KG SPCMA1337-50KGBL
L-Arginine hydrochloride USP
L-Arginine hydrochloride
Formule: C₆H₁₅ClN₄O₂
MW: 210.66 g/mol
Smeltpunt: 218
Dichtheid: 1.42 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064550
CAS nummer: 1119-34-2
EINECS: 214-275-1
Merck Index: 13,00785
Order Now
Specification Test Results
Assay 98.5% Min.