DL-Malic acid NF
Leverancier: Spectrum Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Gevaar
Synoniemen: DL-Hydroxysuccinic acid, (±)-Malic acid, (±)-Hydroxy-1,4-butanedioic acid, (±)-Hydroxysuccinic acid, Hydroxybutanedioic acid, 2-Hydroxysuccinic acid
M1336-12KG M1336-2.5KG M1336-500GM
SPCMM1336-12KGEA 1650 EUR
SPCMM1336-12KG SPCMM1336-2.5KG SPCMM1336-500GM
DL-Malic acid NF
DL-Malic acid
Formule: HOOCCH(OH)CH₂COOH
MW: 134.09 g/mol
Kookpunt: 306
Smeltpunt: 131…132 °C
Dichtheid: 1.601 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064212
CAS nummer: 617-48-1
EINECS: 210-514-9
Merck Index: 13,05730
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=920b54632a869bc5df22624fb9fecd4d11cae65e&vid=278193678126274041defdc0bd71c7c40efb3085&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 99% Min.