4-Aminobenzoic acid USP
Leverancier: Spectrum Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Vitamin Bx, para-Aminobenzoic acid, PABA, Vitamin H1
AM150-50KGBL AM150-100GM AM150-500GM AM150-12KG
SPCMAM150-50KGBLEA 0 EUR
SPCMAM150-50KGBL SPCMAM150-100GM SPCMAM150-500GM SPCMAM150-12KG
4-Aminobenzoic acid USP
4-Aminobenzoic acid
BSE/TSE statement (available upon request)
Allergen statement (available upon request)
Formule: C₇H₇NO₂
MW: 137.14 g/mol
Kookpunt: 340
Smeltpunt: 188
Dichtheid: 1.45 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00007894
CAS nummer: 150-13-0
EINECS: 205-753-0
Merck Index: 13,00422
Order Now
Specification Test Results
Assay 98.5% Min.