Potassium-D-gluconate, anhydrous USP
Leverancier: Spectrum Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Potassium Gluconate, Gluconic Acid Potassium Salt, D-Gluconic acid potassium salt, Potassium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
PO178-12KG PO178-2.5KG PO178-50KGBL
SPCMPO178-12KGEA 1190 EUR
SPCMPO178-12KG SPCMPO178-2.5KG SPCMPO178-50KGBL
Potassium-D-gluconate, anhydrous USP
Potassium-D-gluconate
Formule: HOH₂C(CH(OH))₄COOK
MW: 234.25 g/mol
Smeltpunt: 174…176 °C
Dichtheid: 1.73 g/cm³ (20 °C)
MDL Nummer: MFCD00064211
CAS nummer: 299-27-4
EINECS: 206-074-2
Merck Index: 12,07796
Order Now
Specification Test Results
Assay 97% Min.