Sodium gluconate USP
Leverancier: Spectrum Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: D-Gluconic acid sodium salt, Sodium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconic acid, sodium salt, Sodium D-gluconate
S1286-12KG S1286-2.5KG S1286-500GM
SPCMS1286-12KGEA 966 EUR
SPCMS1286-12KG SPCMS1286-2.5KG SPCMS1286-500GM
Sodium gluconate USP
Sodium gluconate
Formule: HOH₂C(CH(OH))₄COONa
MW: 218.14 g/mol
Smeltpunt: 170…175 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064210
CAS nummer: 527-07-1
EINECS: 208-407-7
Merck Index: 12,08766
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eafd5b786cb23fb5c20dd64ad056ed2d80dc0186&vid=b1ffe48dafca4b8bd9b0d709ea3e986099051767&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 98% Min.