Stearic acid, powder NF
Leverancier: Spectrum Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Octadecanoic acid, Octadecylic acid
ST110-12KG ST110-2.5KG ST110-45KGBL
SPCMST110-12KGEA 1240 EUR
SPCMST110-12KG SPCMST110-2.5KG SPCMST110-45KGBL
Stearic acid, powder NF
Stearic acid
Form: Powder
Formule: C₁₈H₃₆O₂
Kookpunt: 361 °C (1013 hPa)
Smeltpunt: 67 °C
Dichtheid: 0,94 g/cm³ (20 °C)
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00002752
CAS nummer: 57-11-4
EINECS: 200-313-4
Merck Index: 13,08882
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b000f914d1ecf0e3b0d3bc79cd318f3c7842831d&vid=b53ff33f46eb3558f286c08be23286aab54dce6c&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay 40% Min.