Settlometer kit
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
HACH2115700EA 309 EUR
HACH2115700
Settlometer kit
Trechters Sedimentatiekegels
This kit is used for analyzing activated sludge process. Custom PP paddle allows agitation of sample without scratching the jar.
  • Easy-to-read white graduations
  • Impact-resistant
  • Chemically resistant at pH range 3 to 10
Leveringsinformatie: Kit includes a transparent PC 2 liter jar with cover and 10 PC graduated centrifuge tubes (15 ml).