Silica reagents for 5000 silica analyzer
Leverancier: Hach
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
199503 2347003 2353103 2100803 4562700
HACH199503EA 155 EUR
HACH199503 HACH2347003 HACH2353103 HACH2100803 HACH4562700
Silica reagents for 5000 silica analyzer
Milieutestkits
One unit each of the reagents is sufficient for 30 days of operation. The standard solution is sufficient for ten calibrations.