Calcium hydrogen orthophosphate, watervrij, powder USP
Leverancier: Spectrum Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
C1890-12KG C1890-45KGBL
SPCMC1890-12KGEA 1280 EUR
SPCMC1890-12KG SPCMC1890-45KGBL
Calcium hydrogen orthophosphate, watervrij, powder USP
Calcium hydrogen orthophosphate
Form: Powder
Formule: CaHPO₄
Smeltpunt: 370 °C
Dichtheid: 2,89 g/cm³ (20 °C)
MDL Nummer: MFCD00010909
CAS nummer: 7757-93-9
EINECS: 231-826-1
Merck Index: 13,01697
Order Now
Specification Test Results
Assay 98% Min.