Calcium-D-gluconate monohydrate ≥98.5% USP
Leverancier: Spectrum Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: D-Gluconic acid calcium salt monohydrate, Calcium gluconate monohydrate, Gluconic acid calcium salt monohydrate
CA146-12KG CA146-2.5KG CA146-50KGBL
SPCMCA146-12KGEA 1280 EUR
SPCMCA146-12KG SPCMCA146-2.5KG SPCMCA146-50KGBL
Calcium-D-gluconate monohydrate ≥98.5% USP
Calcium-D-gluconate monohydrate
Formule: C₁₂H₂₂CaO₁₄·H₂O
MW: 448.39 g/mol
Smeltpunt: 178
MDL Nummer: MFCD00150787
CAS nummer: 66905-23-5
EINECS: 206-075-8
Order Now
Specification Test Results
Assay 98.5% Min.