Charcoal activated, powder USP
Leverancier: Spectrum Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synoniemen: Carbon activated, Activated carbon, Activated charcoal
CA191-12KG CA191-2.5KG CA191-45KGBL CA191-500GM
SPCMCA191-12KGEA 4400 EUR
SPCMCA191-12KG SPCMCA191-2.5KG SPCMCA191-45KGBL SPCMCA191-500GM
Charcoal activated, powder USP
Charcoal activated
Form: Powder
Formule: C
Smeltpunt: 3550 °C
Dichtheid: 2 g/cm³ (20 °C)
MDL Nummer: MFCD00133992
CAS nummer: 7440-44-0
EINECS: 231-153-3
UN: 1362
ADR: 4.2,III
Bereik: 01-2119488894-16
Merck Index: 13,01818