Pipette tips, wide bore, ART™
Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
2079G 2160G 3591 3701 3592 3702 3590
732-2218EA 77.9 EUR
732-2218 732-2232 732-2335 732-2340 732-2336 732-2341 732-2254
Pipette tips, wide bore, ART™
Pipettips
Transparent PP, sterile or non sterile.
  • Available as non sterile bulk or racked; and as sterile racked
  • Certified sterile tips, free of DNase, RNase, DNA, ATP and pyrogens (sterilised by gamma radiation)
  • Wide bore: Ideal for genomic DNA, fragile cell lines and other viscous materials
Bestelinformatie: Supplied loose bagged and non sterile, sterile in racks.