Sodium polyanetholesulphonate, powder, Reagent Grade

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

0210292390 0210292305 0210292325 0210292380
ICNA0210292390EA 3247 EUR
ICNA0210292390 ICNA0210292305 ICNA0210292325 ICNA0210292380
Sodium polyanetholesulphonate, powder, Reagent Grade
Sodium polyanetholesulphonate

Soluble in water (100 mg/ml, swells and dissolves slowly); insoluble in alcohol.


Polyanetholesulphonic acid sodium salt is a polymer of anetholesulphonic acid.


Polyanetholesulphonic acid sodium salt is used as Anticoagulant.


Polyanetholesufonic acid has effect on antithrombin III activity.

Formule: (C₁₀H₁₁O₄SNa)n
MDL Nummer: MFCD00148427
CAS nummer: 55963-78-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR