Amberlyst® 15 (H), beads
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0215033690 0215033680
ICNA0215033690EA 420 EUR
ICNA0215033690 ICNA0215033680
Amberlyst® 15 (H), beads
Amberlyst® 15 (H)
Soluble in water.
Formule: (C₁₀H₁₀·C₈H₈O₃S)x
MDL Nummer: MFCD00145841
CAS nummer: 9037-24-5