D-ribulose ≥85%, liquid

Leverancier: MP Biomedicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

0215202880 0215202890 0215202810 0215202825
ICNA0215202880EA 457 EUR
ICNA0215202880 ICNA0215202890 ICNA0215202810 ICNA0215202825
D-ribulose ≥85%, liquid
D-ribulose
Formule: C₅H₁₀O₅
CAS nummer: 488-84-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR