(6R)-5,6,7,8-Tetrahydro-L-biopterin dihydrochloride, powder
Leverancier: MP Biomedicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
0215362250 0215362225
ICNA0215362250EA 494 EUR
ICNA0215362250 ICNA0215362225
(6R)-5,6,7,8-Tetrahydro-L-biopterin dihydrochloride, powder
(6R)-5,6,7,8-Tetrahydro-L-biopterin dihydrochloride
Soluble in water (2,3 g completely soluble in 100 ml). A 1 mM solution in water results in a pH of 3,0 and 1 M solution in water results in a pH of 0,45.
Stability: (6R)-BH4.2HCl is hygroscopic. It reacts with oxygen especially in neutral and basic solutions, but dry, in ampules and at −20 °C or colder it can be stored for several years. Oxidised tetrahydrobiopterin solutions become yellow but are relatively stable.
(6R)-5,6,7,8-Tetrahydro-L-biopterin Dihydrochloride is necessary in vivo for the conversion of phenylalanine to tyrosine and for the production of the hormone epinephrine and of the monoamine neurotransmitters, serotonin, dopamine, and norepinephrine. It is also involved in apoptosis and other cellular events mediated by nitric oxide production and catalyses the cleavage of alkylglycerol ethers.
Natural cofactor for phenylalanine hydroxylase, tyrosine hydroxylase, tryptophan hydroxylase, and nitric oxide synthetase.
Formule: C₉H₁₅N₅O₃·2HCl
MDL Nummer: MFCD00891665
CAS nummer: 17528-72-2